Learn More
Enzo Life Sciences TMTH . hydrochloride (50mg)
SDP
Supplier:  Enzo Life Sciences ALX430133M050
Encompass_Preferred
Cyclic hydroxylamine spin probe for in vivo, intraperitoneal and intravenous injections. Lifetime of more than 4 hours in blood plasma, in cells and tissue. Allows quantitative measurements of extracellular O2•– and quantification of intracellular O2•– in cells and tissue samples and in vivo O2•– detection. Alternative name: N-(1-Hydroxy-2,2,6,6-tetramethylpiperidin-4-yl)-2-methylpropanamide . HCl. Purity: ≥99% (EPR). Solubility: Soluble in water, methanol or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-9225
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.