Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Toluene, 99.5% min., ACS, MilliporeSigma™
Shop All MilliporeSigma Products
Click to view available options
Quantity:
500 mL
200 L
Specifications
Specifications
Color | Colorless |
Quantity | 500 mL |
Physical Form | Liquid |
Assay Percent Range | 99.5% min. |
Formula Weight | 92.14 |
Chemical Name or Material | Toluene, For ACS analysis |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction