Learn More
TARGETMOL CHEMICALS INC TONAPOFYLLINE 5MG
Supplier:  TARGETMOL CHEMICALS INC T17117
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Tonapofylline is an orally active and selective antagonist of the A1 adenosine receptor (Ki 7.4 nM for human A1). Tonapofylline can be used in studies about heart failure. purity: 99%
Catalog No. 50-258-1123
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.