Learn More
Restek™ Topaz Drilled Uniliner Inlet Liner with Hole

Features revolutionary technology and inertness to deliver next level True Blue Performance
Supplier: Restek™ 23281
Description
- Deactivation: Unbelievably low breakdown for accurate and precise low-level GC analyses
- Reproducibility: Unbeatable manufacturing controls and QC testing for superior reliability across compound classes
- Productivity: Unparalleled cleanliness for maximized GC uptime and lab throughput
- 100% Satisfaction: If a liner does not perform to expectations, it will be replaced or account will be credited
- Patented
Specifications
PerkinElmer PSS Inlets | |
86.2 mm | |
6.2 mm |
5/Pk. | |
2 mm | |
Topaz |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.