Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Topiramate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. p-7158398 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle

T1696, 97240-79-4, C12H21NO8S

Spectrum™ Chemical Topiramate, USP is used alone or with other medications to prevent and control seizures like epilepsy. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 97240-79-4
Molecular Formula C12H21NO8S
Molecular Weight (g/mol) 339.36
InChI Key KJADKKWYZYXHBB-XBWDGYHZSA-N
IUPAC Name [(1R,2S,6S,9R)-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[7.3.0.0²,⁶]dodecan-6-yl]methyl sulfamate
SMILES CC1(C)O[C@@H]2CO[C@@]3(COS(N)(=O)=O)OC(C)(C)O[C@H]3[C@@H]2O1

Specifications

CAS Max % 1
Ignition Residue 0.002
Packaging Amber Glass Bottle
Quantity 5 g
Specific Rotation -28.6° to -35.0°
Percent Purity 98 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.