Learn More
Agilent Technologies TOTAL INORGANIC CARBON STD
Supplier:  Agilent Technologies ICC033
Encompass
Total inorganic carbon (TIC) standard 1000 mg/LAnalysis Type: QuantitativeAnalyte Count: 2Chemical Type: OrganicConcentration: Sodium bicarbonate (as carbon) 500 g/mL, CAS#:144-55-8 Sodium carbonate (as carbon) 500 g/mL, CAS#:497-19-8Format: Multi component mixtureQuality Standard: ISO 17034Solvent: Water (low TOC, < 50 ppb)Volume: 250 mL
Catalog No. NC2195554
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.