Learn More
ERA WATERS CORP Total Organic Carbon (TOC) - 1000mg/l, 500ml
SDP
Supplier:  ERA WATERS CORP 978
Encompass
One 500ml amber glass bottles with TOC at 1,000mg/l perserved with H2SO4.
Catalog No. NC9477698
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.