Learn More
Enzo Life Sciences ROS-ID Total ROS detection kit (1x1Kit)
Alternative Name: Reactive oxygen species. Applications: Flow Cytometry. Fluorescence microscopy. Fluorescent detection, HTS. Contents: Oxidative Stress Detection Reagent (Green), 300 nmoles . ROS Inducer (Pyocyanin), 1 µmole. ROS Inhibitor (N-acetyl-L-cysteine), 2 x 10 mg. Wash Buffer Salts, 1 pack