Learn More
Enzo Life Sciences ROS-ID Total ROS/Superoxide detection kit (1x1Kit)
SDP
Supplier:  Enzo Life Sciences ENZ51010
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Directly Monitor Global Levels  of Reactive Oxygen Species (ROS), and Specifically Superoxide. Applications: Flow Cytometry. Fluorescence microscopy. Fluorescent detection, HTS. Contents: Oxidative Stress Detection Reagent (Green), 300 nmoles. Superoxide Detection Reagent (Orange), 300 nmoles . ROS Inducer (Pyocyanin), 1 µmole. ROS Inhibitor (N-acetyl-L-cysteine), 2 x 10 mg. Wash Buffer Salts, 1 pack
Catalog No. NC9903488