Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Toyocamycin
Adenosine analog; antifungal antibiotic
$85.70 - $361.50
Specifications
Purity | >98% |
---|---|
Product Type | Toyocamycin |
Molecular Weight (g/mol) | 291.26 |
Inhibitors | Toyocamycin |
Description
Adenosine analog; antifungal antibiotic. Inhibits RNA self-cleavage in HEK79 cells and PI 3-kinase activity in A431 epidermoid carcinoma cell membrane fractions. Also inhibits thapsigargin-induced XBP1-luciferase activation and induces apoptosis in multiple myeloma cell lines.Specifications
>98% | |
Toyocamycin | |
291.26 | |
Toyocamycin |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title