Learn More
Trace Metals Standards without Mercury For Environmental Analysis, Set of 3, SPEX CertiPrep™
Shop All Spex CertiPrep Products

Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
- Trace Metals
- 125 mL volume
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Chemical Name or Material
Trace Metals without Mercury
Name Note
Set of 3 Groundwater and Wastewater Pollution Control Check Standard
Packaging
Triple-leached LDPE bottle
Quantity
3 x 125 mL
Concentration or Composition Notes
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment)
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Grade
Environmental
Product intended for laboratory use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.