Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tracazolate hydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
GABAA receptor modulator. Can potentiate or inhibit recombinant GABAA function depending on subunit combination. Enhances native GABAA receptor function and is anxiolytic following systemic administration in vivo.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 340.85 |
Percent Purity | >99% |
Chemical Name or Material | Tracazolate hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction