Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Dopamine hydrochloride certified reference material, MilliporeSigma™ Supelco™

Catalog No. 111018687 Shop All MilliporeSigma Supelco Products
Click to view available options
:
50 mg

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Specifications

Quantity 50 mg
CAS 62-31-7
Type Certified Reference Material
Product Line TraceCERT
Linear Formula (HO)2C6H3 CH2 CH2 NH2·HCl
MDL Number MFCD00012898
Synonym 2-(3,4-Dihydroxyphenyl)ethylamine hydrochloride; 3,4-Dihydroxyphenethylamine hydrochloride; 3-Hydroxytyramine hydrochloride; 4-(2-Aminoethyl)-1,2-benzenediol hydrochloride
Formula Weight 189.64
Grade TraceCERT™ Analytical Standard
Melting Point 248°C to 250°C
Physical Form Powder
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.