Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TraceCERT™ Ethylene carbonate Standard, MilliporeSigma™ Supelco™

Catalog No. 111015978 Shop All MilliporeSigma Supelco Products
Click to view available options
:
1 g

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Cleaning Products, Cosmetics, Flavors and Fragrances, Food and Beverages and Personal Care

Specifications

Quantity 1 g
CAS 96-49-1
Type Certified Reference Material
Product Line TraceCERT
Linear Formula C3H4O3
MDL Number MFCD00005382
Synonym 1,3-Dioxolan-2-one
Formula Weight 88.06
Grade TraceCERT™ Analytical Standard
Vapor Pressure 0.02 mmHg (36.4°C)
Vapor Density 3.04 (vs. air)
Density 1.321 g/mL (at 25°C)
Melting Point 35°C to 38°C
Boiling Point 243°C to 244°C (740 mmHg)
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.