Learn More
Enzo Life Sciences trans-7-Hydroxy-PIPAT. maleate (5mg). CAS: 159559-71-4
SDP
Supplier:  Enzo Life Sciences BMLD1060005
Encompass_Preferred
A selective, high affinity D3 dopamine receptor ligand (Ki=0.99 nM). Alternative name: trans-7-Hydroxy-2-[N-propyl-N(3'-iodo-2'-propenyl)amino]tetralin . maleate. Purity: ≥98%. Long Term Storage: -20°C.
Catalog No. 50-201-0492
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.