Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans,trans-1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one 98.0+%, TCI America™
SDP

Catalog No. p-7123560 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 103836-71-1
Molecular Formula C19H12F6O
Molecular Weight (g/mol) 370.294
InChI Key OIUBVYQZMUKRRF-YDWXAUTNSA-N
Synonym trans,trans-Bis[4-(trifluoromethyl)benzal]acetone, trans,trans-Bis[4-(trifluoromethyl)benzylidene]acetone, trans,trans-Bis[4-(trifluoromethyl)styryl] Ketone
PubChem CID 5373276
IUPAC Name (1E,4E)-1,5-bis[4-(trifluoromethyl)phenyl]penta-1,4-dien-3-one
SMILES C1=CC(=CC=C1C=CC(=O)C=CC2=CC=C(C=C2)C(F)(F)F)C(F)(F)F

Specifications

Quantity 200 mg
Formula Weight 370.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-1,5-Bis[4-(trifluoromethyl)phenyl]-1,4-pentadien-3-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.