Learn More
Gold Biotechnology Inc trans-Zeatin 50 mg
SDP
Supplier:  Gold Biotechnology Inc Z10550
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
trans-Zeatin is a naturally occurring cytokinin plant growth regulator routinely used in plant tissue culture to induce cell division and shoot formation. trans-zeatin is freely soluble in acetic acid and water.
Catalog No. NC2943394