Learn More
Gold Biotechnology Inc trans-Zeatin-riboside 250 mg
Supplier: Gold Biotechnology Inc Z100250
Zeatin Riboside is the most active and ubiquitous form of the naturally occurring cytokinins. Cytokinins promote cell division, stimulate shoot proliferation, inhibit root formation, slow the aging process, and activate gene expression and metabolic activity in general. In mature plants, natural cytokinin hormone levels are high during maximum growth periods. Concentrations of cytokinin used for horticulture vary between 0.1 to 10 mg/L.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.