Learn More
Cole-Parmer Ashcroft K1150PSI Thin Film Pressure Transmitter, 0 to 150 psi, 1.0%
Supplier:  Cole-Parmer 6844037
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
This transmitter offers the benefits of polysilicon thin film performance at affordable prices. Using chemical vapor deposition methods polysilicon strain gauges are molecularly bonded to a metal diaphragm. This sensor, combined with the latest surface mount electronics promote superior performance in a durable package.
Catalog No. 50-992-544