Learn More
Kamiya Biomedical Transferrin Reagent, 60mL+20mL/PK RX
Supplier:  Kamiya Biomedical KAI023
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
K-ASSAY Transferrin Assay Reagent. R1: 3 x 20mL (liquid), R2: 2 x 10mL (liquid). Assay Range: 45 - 455 mg/dL. The K-ASSAY Transferrin assay is intended for the quantitative determination of human transferrin in human serum by immunoturbidimetric assay. FOR IN VITRO DIAGNOSTIC USE. Required Calibrator: K-ASSAY Multi-Analyte Calibrator (Cat. No. KAI-016C). Recommended Controls: K-ASSAY Liquid ITA Control (Cat. No. K49C-2M or K49C-4M). This assay is intended for use on most popular chemistry analyzers that allow open channel reagents. Please contact KAMIYA BIOMEDICAL COMPANY to check parameter availability for your analyzer.
Catalog No. NC0909191
missing translation for 'QuantityLeft'