Learn More
StatLab TRANSFORM SPAY 1 PINT
SDP
Supplier:  StatLab CWTFSPT
Encompass_Preferred
TransForm Spray destroys formalin and its fumes in a little as three seconds. Just spray directly onto grossing area, sinks, countertops, or anywhere formalin fumes are a problem.
Catalog No. NC9532498
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.