Learn More
CUSABIO BIOTECHNOLOGY LLC Human Transforming Growth factor β1,TGF-β1 ELISA kit
Supplier: CUSABIO BIOTECHNOLOGY LLC CSBE04725HIS
This Human Transforming Growth factor β1,TGF-β1 ELISA kit was designed for the quantitative measurement of Human Transforming Growth factor β1 protein in serum, plasma, cell culture supernates. It is a Sandwich ELISA kit, its detection range is 23.5 pg/ml-1500 pg/ml and the sensitivity is 5.8 pg/ml. One kit contains 96 wells.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.