Learn More
Corning™ Transforming Growth Factor-b (TGF-b), Human Natural

Corning™ Transforming Growth Factor-beta (TGF-beta), human natural is a multifunctional protein that plays a central role in the regulation of cell growth and differentiation with either stimulatory or inhibitory effects.
$1144.00
Specifications
Content And Storage | ≥ 3 months from date of shipment (at -70°C) |
---|---|
Purity or Quality Grade | >95% by SDS-PAGE |
Source | Human Platelets |
Description
Formulation
Frozen in 36% acetonitrile-0.1% trifluoroacetic acid
Caution: TGF-ß absorbs to glass and plastic surfaces (instructions for dilution are provided with the product)
Source
Human platelets. NOTE: Source material tested for hepatitis B antigen and HIV-1 antibody
Molecular Weight
25 kD
Quality
- Purity: >95% by SDS-PAGE
- Tested for ability to inhibit DNA synthesis of Mv 1Lu mink lung cells (ATCC CCL-64)
- Tested and found negative for bacteria, fungi and mycoplasma
Tip
Recommended concentration: 10-50 pg/mL of medium depending on cell type and application.
Preparation and Storage
Stable for at least three months at -70°C (if diluted according to instructions)
Storage
Shelf life: ≥ 3 months from date of shipment at -70°C
Specifications
≥ 3 months from date of shipment (at -70°C) | |
>95% by SDS-PAGE | |
Human Platelets |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.