Learn More
Kyfora Bio Transporter 5 AmpliFect Tra 1
Supplier:  Kyfora Bio 144151
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Kyfora Bio's Transporter 5 + AmpliFect transfection kit contains our trusted, highly efficient PEI transfection reagent, Transporter 5, as well as our cutting-edge transfection enhancer, AmpliFect. AmpliFect is designed to work synergistically with Transporter 5 transfection workflows resulting in a 2x increase in viral vector (AAV, LV), recombinant protein, and mAb titers. When used in combination, Transporter 5 and AmpliFect offer superior performance that increases productivity and reduces manufacturing costs compared to other transfection solutions on the market.
Catalog No. 50-274-3943