Learn More
Kamiya Biomedical TRAP (Tartrate-resistant acidic phosphatase) STAINING KIT
Supplier:  Kamiya Biomedical KT008
Encompass_Preferred
TRAP (Tartrate-resistant acidic phosphatase) STAINING KIT. For the staining of tartrate-resistant acid phosphatase in osteoclasts.
Catalog No. NC1723513
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.