Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-n-propyltin chloride, 95%, Thermo Scientific Chemicals

Catalog No. p-7051533
Click to view available options
Quantity:
1 g
5 g
25 g

Tri-n-propyltin chloride is used as pharmaceutical intermediate and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2279-76-7
Molecular Formula C9H21ClSn
Molecular Weight (g/mol) 283.427
MDL Number MFCD00013610
InChI Key FVFLIQMADUVDSP-UHFFFAOYSA-M
Synonym tri-n-propyltin chloride, tripropyltin chloride, stannane, chlorotripropyl, tripropylstannium chloride, tin, tripropyl-, chloride, unii-7iq49g05u7, chloro tripropyl stannane, tri-n-propyl tin chloride, chlorotripropyl stannane, tri-n-propyltinchloride
PubChem CID 16784
IUPAC Name chloro(tripropyl)stannane
SMILES CCC[Sn](CCC)(CCC)Cl

Specifications

Melting Point -23.5°C
Density 1.2678 g/mL at 28°C
Boiling Point 123°C (13 mmHg)
Refractive Index 1.491
Quantity 1 g
UN Number UN2788
Sensitivity Moisture sensitive
Solubility Information Sparingly Soluble in water (4.0E-3g/L) (25°C).
Formula Weight 283.41
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tri-n-propyltin chloride
Show More Show Less
Hazard Category H301+H311+H331
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
EINECSNumber 218-910-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.