Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-o-tolyl Phosphite 97.0+%, TCI America™
SDP

Catalog No. p-7135126 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 2622-08-4
Molecular Formula C21H21O3P
Molecular Weight (g/mol) 352.37
MDL Number MFCD00014911
InChI Key BKHZQJRTFNFCTG-UHFFFAOYSA-N
Synonym tri-o-tolyl phosphite, tri-o-tolylphosphite, tri-o-cresyl phosphite, phosphorous acid tri-o-cresyl ester, tris o-methylphenyl phosphite, tris 2-methylphenyl phosphite, tris 2-tolyl phosphite, tris o-tolyloxy phosphine, phosphorous acid, tri-o-cresyl ester, tri-2-tolylphosphite
PubChem CID 92937
IUPAC Name tris(2-methylphenyl) phosphite
SMILES CC1=CC=CC=C1OP(OC1=CC=CC=C1C)OC1=CC=CC=C1C

Specifications

Color Yellow
Boiling Point 238°C
Quantity 1 g
Formula Weight 352.37
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tri-o-tolyl Phosphite
EINECSNumber (3)-2510
RTECSNumber TH1050000
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.