Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-p-tolyl Phosphite, TCI America™
SDP

Catalog No. P117525G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 620-42-8
Molecular Formula C21H21O3P
Molecular Weight (g/mol) 352.37
MDL Number MFCD00041909
InChI Key FEVFLQDDNUQKRY-UHFFFAOYSA-N
Synonym tri-p-tolyl phosphite, tri-p-cresyl phosphite, tris p-tolyl phosphite, tris p-methylphenyl phosphite, tritolyl phosphite, phosphorous acid, tris 4-methylphenyl ester, p-tolyl phosphite c7h7o 3p, tris 4-methylphenyl phosphite, phosphorous acid, tri-p-tolyl ester, tris methylphenyl phosphite
PubChem CID 12106
IUPAC Name tris(4-methylphenyl) phosphite
SMILES CC1=CC=C(OP(OC2=CC=C(C)C=C2)OC2=CC=C(C)C=C2)C=C1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 352.37
Physical Form Liquid
Chemical Name or Material Tri-p-tolyl Phosphite
RTECSNumber TH1100000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.