Learn More
Alkali Scientific Triangle Spreader, 30 x 207mm (25/Sterile bag, 4 Bags), 100/cs
SDP
Supplier:  Alkali Scientific TS5451
Encompass_Preferred
These triangle shape spreaders are also used for CFU count applications, but have a grooved handle that improves grip and orientation. Made of polypropylene, they are available with a 30mm span.
Catalog No. 50-238-8818
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.