Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyrin, MP Biomedicals™

Catalog No. p-7083108 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
500 mL
1L

Used as a deliverable source of natural butyrate. Tributyrin, MP Biomedicals is glycerol substituted with 3 butyrl esters. It induces differentiation of malignant cells, followed by arrest of cell growth and apoptosis. It potently inhibits growth of and i

Chemical Identifiers

CAS 60-01-5
Molecular Formula C15H26O6
Molecular Weight (g/mol) 302.367
InChI Key UYXTWWCETRIEDR-UHFFFAOYSA-N
Synonym tributyrin, glyceryl tributyrate, glycerol tributyrate, tributin, butyrin, butyryl triglyceride, propane-1,2,3-triyl tributyrate, glycerol tributanoate, glyceroltributyrin, tri-n-butyrin
PubChem CID 6050
ChEBI CHEBI:35020
IUPAC Name 2,3-di(butanoyloxy)propyl butanoate
SMILES CCCC(=O)OCC(COC(=O)CCC)OC(=O)CCC

Specifications

Melting Point -75°C (literature)
Density 1.032g/mL (Lit.)
Boiling Point 287°C to 288°C (literature)
Refractive Index n20/D 1.435 (Lit.)
Quantity 500 mL
Solubility Information Soluble in alcohol and ether; insoluble in water.
Formula Weight 302.36
Percent Purity ≥96%
Physical Form Liquid
Recommended Storage Room Temperature (15–30°C).
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.