Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic acid, 99.6%, MP Biomedicals™

Catalog No. ICN19052180 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

76-03-9

Molecular Formula

C2HCl3O2

Molecular Weight (g/mol)

163.378

InChI Key

YNJBWRMUSHSURL-UHFFFAOYSA-N

Synonym

trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer

PubChem CID

6421

ChEBI

CHEBI:30956

IUPAC Name

2,2,2-trichloroacetic acid

SMILES

C(=O)(C(Cl)(Cl)Cl)O

Specifications

Color

White

Assay Percent Range

0.996

Formula Weight

163.4

Density

1.61g/cm3

Quantity

100 g

Physical Form

Crystalline Powder

Chemical Name or Material

Trichloroacetic acid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.