Learn More
Active Motif Trichostatin A
SDP
Supplier:  Active Motif 14039
Encompass_Preferred
Active Motif's Trichostatin A is a potent and selective histone deacetylase (HDAC) inhibitor (Ki = 3.4 nM). It induces reversion of ras-transformed cells to normal morphology and induces dedifferentiation of primordial germ cells into embryonic germ cells. It is cell permeable, and is active in vivo.
Catalog No. NC1838553
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.