Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Triethylamine, BAKER ANALYZED™ Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Specifications
Specifications
Flash Point | −11.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Quantity | 500mL |
Specific Gravity | 1L = 0.73kg |
Formula Weight | 101.19 |
Grade | BAKER ANALYZED™ Reagent |
Chemical Name or Material | Triethylamine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction