Learn More
TARGETMOL CHEMICALS INC TRIFLUOPERAZINE 10MG
Supplier:  TARGETMOL CHEMICALS INC T8389
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Trifluoperazine (trifluoroperazine) is a Dopamine D2 receptor inhibitor(IC50 1.2 nM) and Treatment of schizophrenia. purity: 99%
Catalog No. 50-257-6244
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.