Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trifluoroacetic Acid, HPLC Grade, J.T.Baker™
Shop All Avantor Products
Click to view available options
Quantity:
10 x 1mL
500mL
900mL
Packaging:
Glass Ampoule
Narrow Mouth Amber Glass Bottle
Specifications
Specifications
Formula Weight | 114.03 |
Grade | HPLC |
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 1.53kg |
Quantity | 500mL |
Chemical Name or Material | Trifluoroacetic Acid, Gradient Tested for HPLC Peptide and Protein Analysis |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction