Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic Acid, LC-MS Ultra, Honeywell™ Fluka™

Catalog No. p-7120894 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 mL
2 mL
50 mL
Packaging:
Ampoule
Glass Bottle

LC-MS Ultra, Eluent additive for UHPLC-MS

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise
  • Exemplary lot-to-lot reproducibility
  • Low levels of trace metals
  • Packaged in clear borosilicate glass bottles

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
SMILES OC(=O)C(F)(F)F

Specifications

Melting Point -15°C
Boiling Point 72°C
Color Colorless
Linear Formula CF3COOH
UN Number UN2699
Beilstein 742035
Solubility Information Soluble in water
IUPAC Name trifluoroacetic acid
Formula Weight 114.02g/mol
Grade LC-MS Ultra
Density 1.489g/mL at 20°C
Quantity 1 mL
Flash Point >100°C (>212°F)
Packaging Ampoule
Physical Form Liquid
Refractive Index n20/D 1.3(lit.)
Vapor Density 3.9 (vs air)
Vapor Pressure 97.5mmHg (20°C)
Chemical Name or Material Trifluoroacetic Acid
Show More Show Less
Hazard Statement P273-P280-P305 + P351 + P338-P310
Precautionary Statement H314-H332-H412
DOTInformation 8/PGI
EINECSNumber 200-929-3
RTECSNumber AJ9625000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.