Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluorothymidine, 98%, Thermo Scientific Chemicals

Catalog No. p-4813697
Click to view available options
Quantity:
250 mg
1 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70-00-8
Molecular Formula C10H11F3N2O5
Molecular Weight (g/mol) 296.20
MDL Number MFCD00006534
InChI Key VSQQQLOSPVPRAZ-RRKCRQDMSA-N
Synonym trifluridine, trifluorothymidine, viroptic, 5-trifluorothymidine, trifluridina, trifluridinum, virophta, trifluoromethyldeoxyuridine, f3tdr, f3dthd
PubChem CID 6256
ChEBI CHEBI:75179
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(trifluoromethyl)pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C(=O)NC1=O)C(F)(F)F

Specifications

Melting Point 190°C to 193°C
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Merck Index 159855
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in isopropanol
Formula Weight 296.2
Percent Purity 98%
Chemical Name or Material Trifluorothymidine
Hazard Category Carcinogenicity (category 2), Acute toxicity (category 4)
Hazard Statement Suspected of causing cancer, Harmful if swallowed, in contact with skin or if inhaled
EINECSNumber 200-722-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.