Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trilaurin 98.0+%, TCI America™
SDP

Catalog No. p-7130137 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
100 g
500 g

Chemical Identifiers

CAS 538-24-9
Molecular Formula C39H74O6
Molecular Weight (g/mol) 639.015
MDL Number MFCD00026559
InChI Key VMPHSYLJUKZBJJ-UHFFFAOYSA-N
Synonym trilaurin, glycerol trilaurate, glyceryl tridodecanoate, glyceryl trilaurate, glycerin trilaurate, lauric acid triglyceride, tridodecanoin, lauric acid triglycerin ester, trilauroylglycerol, laurin, tri
PubChem CID 10851
ChEBI CHEBI:77389
IUPAC Name 2,3-di(dodecanoyloxy)propyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC

Specifications

Melting Point 47°C
Color White-Yellow
Quantity 25 g
Formula Weight 639.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Trilaurin
EINECSNumber (2)-0669
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.