Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl(4-pyridyl)tin 97.0+%, TCI America™
SDP

Catalog No. p-7137026 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 59020-06-3
Molecular Formula C8H13NSn
Molecular Weight (g/mol) 241.91
MDL Number MFCD03844808
InChI Key YCBWXJZXFUKDPU-UHFFFAOYSA-N
Synonym 4-Trimethylstannylpyridine
PubChem CID 11128458
IUPAC Name 4-(trimethylstannyl)pyridine
SMILES C[Sn](C)(C)C1=CC=NC=C1

Specifications

Color Yellow
Quantity 1 g
UN Number 2788
Formula Weight 241.91
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Trimethyl(4-pyridyl)tin
TSCA No
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.