Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylsulfonium Bromide 98.0+%, TCI America™
SDP

Catalog No. p-7136952 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
100 g

Chemical Identifiers

CAS 3084-53-5
Molecular Formula C3H9BrS
Molecular Weight (g/mol) 157.069
MDL Number MFCD00051367
InChI Key GOTIICCWNAPLMN-UHFFFAOYSA-M
Synonym trimethylsulfonium bromide, trimethylsulfoniumbromide, sulfonium, trimethyl-, bromide, trimethylsulfanium bromide, unii-j59ncr7385, sulfonium, trimethyl-, bromide 1:1, acmc-209hj3, ksc492m3f
PubChem CID 2735154
IUPAC Name trimethylsulfanium;bromide
SMILES C[S+](C)C.[Br-]

Specifications

Color White
Quantity 25 g
Formula Weight 157.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylsulfonium Bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.