Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyltin chloride, Thermo Scientific Chemicals

Catalog No. p-7080330
Click to view available options
Quantity:
2 g
10 g
50 g

Trimethyltin chloride is used as a heat stabilizer in the manufacture of vinyl window blinds. It is also used as a stabilizer for plastics and as additive to paint, as antifouling agent. It also serves as a byproduct of methyltin manufacture. Further, it is used as a precursor to vinyltrimethylstannane and in polyvinyl chloride stabilization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1066-45-1
Molecular Formula C3H11ClSn
Molecular Weight (g/mol) 201.28
MDL Number MFCD00000520
InChI Key FVFGWISLDLUGHX-UHFFFAOYSA-N
Synonym trimethyltin chloride, stannane, chlorotrimethyl, chlorotrimethyltin, trimethylchlorotin, trimethylstannyl chloride, trimethylchlorostannane, chloro trimethyl stannane, m&t chemicals 1222-45, stannylium, trimethyl-, chloride, trimethyltinchloride
PubChem CID 14016
IUPAC Name chloro(trimethyl)stannane
SMILES Cl.C[SnH](C)C

Specifications

Melting Point 37°C to 39°C
Color White
Boiling Point 154°C
Flash Point 97°C (207°F)
Odor Unpleasant
Quantity 2 g
UN Number UN3146
Sensitivity Moisture sensitive
Solubility Information Soluble in water,chloroform and organic solvents.
Formula Weight 199.25
Physical Form Crystalline
Chemical Name or Material Trimethyltin chloride
Show More Show Less
Hazard Category H300+H310+H330-H335-H336-H373
Hazard Statement GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
EINECSNumber 213-917-8
RTECSNumber WH6850000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.