Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Triphenyl Phosphate 99.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
25 g
500 g
Specifications
Specifications
Melting Point | 50°C |
Color | White |
Boiling Point | 220°C |
Quantity | 25 g |
UN Number | 3077 |
Formula Weight | 326.29 |
Percent Purity | ≥99.0% (GC) |
Physical Form | Flakes |
Chemical Name or Material | Triphenyl Phosphate |
Safety and Handling
EINECSNumber | (3)-2522 |
RTECSNumber | TC8400000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction