Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylantimony Oxide 95.0+%, TCI America™
SDP

Catalog No. p-7136984 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 4756-75-6
Molecular Formula C18H15OSb
Molecular Weight (g/mol) 369.08
MDL Number MFCD00013837
InChI Key RIBJLXBLWQKWLO-UHFFFAOYSA-N
Synonym Triphenylstibine Oxide
PubChem CID 78497
IUPAC Name triphenylstibanone
SMILES O=[Sb](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 1 g
UN Number 3467
Formula Weight 369.08
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylantimony Oxide
RTECSNumber WJ0900000
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.