Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triphenylsulfonium Bromide 98.0+%, TCI America™
SDP

Catalog No. p-7136871 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g
5 g

Chemical Identifiers

CAS 3353-89-7
Molecular Formula C18H15BrS
Molecular Weight (g/mol) 343.282
MDL Number MFCD00428762
InChI Key VMJFYMAHEGJHFH-UHFFFAOYSA-M
PubChem CID 11984010
IUPAC Name triphenylsulfanium;bromide
SMILES C1=CC=C(C=C1)[S+](C2=CC=CC=C2)C3=CC=CC=C3.[Br-]

Specifications

Melting Point 293°C
Color White
Quantity 100 mg
Formula Weight 343.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triphenylsulfonium Bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.