Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tripyrrolidinophosphine Oxide 98.0+%, TCI America™
SDP

Catalog No. p-7137496 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 6415-07-2
Molecular Formula C12H24N3OP
Molecular Weight (g/mol) 257.32
MDL Number MFCD00014095
InChI Key GQOGEHIVQIMJMO-UHFFFAOYSA-N
Synonym Tris(N,N-tetramethylene)phosphoric Triamide
PubChem CID 80866
IUPAC Name 1-dipyrrolidin-1-ylphosphorylpyrrolidine
SMILES C1CCN(C1)P(=O)(N2CCCC2)N3CCCC3

Specifications

Color Yellow
Boiling Point 133°C
Quantity 5 g
Formula Weight 257.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tripyrrolidinophosphine Oxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.