Learn More
Tris(1-methoxy-2-methyl-2-propoxy)bismuth, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Tris(1-methoxy-2-methyl-2-propoxy)bismuth is used as metal organic precursors to prepare bismuth titanium oxide thin films, which finds application in ferroelectric random access memory.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Specifications
Melting Point | 70°C |
Color | Yellow |
Quantity | 1 g |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in ethylcyclohexane. |
Formula Weight | 518.4 |
Percent Purity | 99.99% |
Physical Form | Crystalline |
Assay | (metals basis) |
Chemical Name or Material | Tris (1-methoxy-2-methyl-2-propoxy)bismuth |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
storageNote1 |
|
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.