Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2-butoxyethyl) Phosphate 95.0+%, TCI America™
SDP

Catalog No. p-7134727 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 78-51-3
Molecular Formula C18H39O7P
Molecular Weight (g/mol) 398.48
MDL Number MFCD00009456
InChI Key WTLBZVNBAKMVDP-UHFFFAOYSA-N
Synonym tris 2-butoxyethyl phosphate, tbep, phosflex t-bep, tris butoxyethyl phosphate, tributoxyethyl phosphate, tri butoxyethyl phosphate, kronitex kp-140, tributyl cellosolve phosphate, ethanol, 2-butoxy-, phosphate 3:1, tri 2-butoxyethyl phosphate
PubChem CID 6540
ChEBI CHEBI:35038
IUPAC Name tris(2-butoxyethyl) phosphate
SMILES CCCCOCCOP(=O)(OCCOCCCC)OCCOCCCC

Specifications

Color Yellow
Boiling Point 220°C
Quantity 25 g
Formula Weight 398.48
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(2-butoxyethyl) Phosphate
EINECSNumber (2)-2022
RTECSNumber KJ9800000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.