Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(4-chlorophenyl) Borate 97.0+%, TCI America™
SDP

Catalog No. B16445G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 7359-58-2
Molecular Formula C18H12BCl3O3
Molecular Weight (g/mol) 393.451
MDL Number MFCD00191614
InChI Key JOAZIIBFQMIXJM-UHFFFAOYSA-N
Synonym Boric Acid Tris(4-chlorophenyl) Ester
PubChem CID 633786
IUPAC Name tris(4-chlorophenyl) borate
SMILES B(OC1=CC=C(C=C1)Cl)(OC2=CC=C(C=C2)Cl)OC3=CC=C(C=C3)Cl

Specifications

Color Yellow
Quantity 5 g
Formula Weight 393.45
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Tris(4-chlorophenyl) Borate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.