Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIS Hydrochloride, Made from Tromethamine USP, Biotech Reagent Grade, J.T.Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly bottle
Specifications
Specifications
Melting Point | 147°C to 153°C |
Packaging | Poly bottle |
Quantity | 500g |
Specific Gravity | 1L = 0.00kg |
Formula Weight | 157.6 |
Grade | GMP manufactured product |
Chemical Name or Material | TRIS Hydrochloride, Made from Tromethamine USP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction