Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRO 19622, Tocris Bioscience™

Catalog No. p-4784722 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Binds voltage-dependent anion channel (VDAC)

Neuroprotective and neuroregenerative compound. Rescues motor neurons from axotomy-induced cell death and promotes nerve regeneration following sciatic nerve crush in vivo. Binds directly to two components of the mitochondrial permeability pore, the voltage-dependent anion channel (VDAC) and translocator protein; thought to inhibit MPTP opening.

Chemical Identifiers

CAS 22033-87-0
Molecular Formula C27H45NO
Molecular Weight (g/mol) 399.663
InChI Key QNTASHOAVRSLMD-GYKMGIIDSA-N
Synonym olesoxime, cholest-4-en-3-one, oxime
PubChem CID 76971721
IUPAC Name N-[(8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-ylidene]hydroxylamine
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4=CC(=NO)CCC34C)C

Specifications

Quantity 10 mg
Formula Weight 399.65
Percent Purity >98%
Chemical Name or Material TRO 19622
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.